Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prolactin, Human Pituitary, Iodination Grade, Protein

Catalog No. 8690395UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-903-95UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 86-903-95UG Supplier MilliporeSigma™ Supplier No. 8690395UG
Add to Cart
Add to Cart
Native, human prolactin from pituitary gland. Single chain peptide hormone required for lactation in mammals. Prolactin receptors are present in breast tissue, adrenals, ovary, testis, kidney, and liver. Potency: ≥30IU/mg (WHO 3rd IS 84/500).
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Acidified EtOH or MeOH
Purification Method By weight
Quantity 5 μg
Immunogen Prolactin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Solid
Purity or Quality Grade ≥98%, Iodination Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.