Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Protein A, Staphylococcus aureus, Protein

Catalog No. 5392025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-920-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-920-25MG Supplier MilliporeSigma™ Supplier No. 5392025MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native protein A from Staphylococcus aureus. Provides a versatile and economical means for isolating, detecting, and purifying antibodies. It is especially useful in immunohistochemical studies because, unlike secondary antibodies, it does not bind to cell Fc receptors and, therefore, will not give high background reactions. Strongly reactive with guinea pig; human IgG1, IgG2, and IgG4; mouse IgG2b, and rabbit. Weakly reactive with bovine; human IgA, IgE, and IgM; and mouse IgG3. Nonreactive with goat; horse; human IgD and IgG3; rat; mouse IgG1; and sheep.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Form Lyophilized Solid
Regulatory Status RUO
Content And Storage Following initial thaw, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.