Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Protein G, Recombinant Protein, E. coli

Catalog No. 5393031MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-930-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-930-31MG Supplier MilliporeSigma™ Supplier No. 5393031MG
Engineered to eliminate non-specific binding with serum albumin.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method H2O (5mg/mL) or Aqueous Buffers (5mg/mL).
Purification Method SDS-PAGE
Quantity 1 mg
Source E. coli
Immunogen Protein G
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Avoid freeze/thaw cycles. Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.