Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pSTBlue-1 Perfectly Blunt Cloning Kit

Catalog No. 701913 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-191-3 20 Reactions
70-191-4 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-191-3 Supplier MilliporeSigma™ Supplier No. 701913
Add to Cart
Add to Cart
The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at _70°C. Avoid freeze/thaw cycles.
Quantity 20 Reactions
For Use With (Application) Simplified cloning of DNA generated by PCR using any type of DNA polymerase

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.