Learn More
MilliporeSigma™ pT7Blue Perfectly Blunt Cloning Kit
Catalog No. 701893 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-189-3 20 Reactions
70-189-4 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-189-3 Supplier MilliporeSigma™ Supplier No. 701893
Add to Cart
Add to Cart
The pT7Blue-3 vector is multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, a T7 promoter and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned at each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 20 Reactions
Vector pT7Blue Perfectly Blunt Vector