Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PTP, Yersinia enterocolitica, Recombinant Protein, E. coli

Catalog No. 5397342000U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-973-42000U 2000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-973-42000U Supplier MilliporeSigma™ Supplier No. 5397342000U
Recombinant, 51kDa catalytic domain of the Yersinia protein tyrosine phosphatase containing a C235R mutation and expressed in E. coli. Suitable for dephosphorylation of phosphotyrosine residues in proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM HEPES, 5mM DTT, 2mM EDTA, 0.01% BRIJ™ 35 Detergent, 50% glycerol, pH 7.0.
Purification Method SDS-PAGE
Quantity 2000 U
Source E. coli
Immunogen PTP
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Y. enterocolitica
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.