Learn More
MilliporeSigma™ PureProteome™ Carboxy FlexiBind Magnetic Bead System
Shop All MilliporeSigma ProductsDescription
Custom affinity bead generation for binding to target ligand.
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Research Use Only
Specifications
Specifications
| Product Type | Carboxy flexiBind Magnetic bead system |
| Quantity | 2 x 1mL |
| Particle Size | 0.3μm |
| Content And Storage | 2° to 8°C |
| For Use With (Application) | Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
| Matrix | Magnetic Beads |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.