Learn More
Description
The PureProteome Protein A/G Mix Magnetic Bead System provides researchers with the power of the combination of both Protein A and Protein G immunoglobulin binding affinities in one convenient magnetic bead product, resulting in greater antibody compatibility than either Protein A or Protein G alone.
- Suitable for immunoprecipitation, crosslinking immunoprecipitation, antibody purification
- Binding capacity: 2–3 mg of Rabbit IgG per mL of suspension
- High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
- Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Fast processing time: Beads are immobilized in seconds; increased kinetics of bead-protein binding enables shorter incubations
- Particle Size: 10μm
Specifications
Specifications
| Product Type | Protein A/G mix Magnetic bead |
| Quantity | 10 mL |
| Particle Size | 10μm |
| Content And Storage | 2° to 8°C |
| For Use With (Application) | Immunoprecipitation, cross-linking immunoprecipitation, Antibody purification |
| Matrix | Magnetic Beads |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.