Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PvuRts1I (5-hmC Restriction Endonuclease)

Catalog No. 141058 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-105-8 200 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-105-8 Supplier MilliporeSigma™ Supplier No. 141058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PvuRts1I is an enzyme that can specifically digest DNA containing 5-hydroxymethylcytosine (5-hmC). This restriction endonucleaseis used to analyze 5-hmC patterns genomic DNA by cleaving both glucosylated and non-glucosylated 5-hmC DNA sequences.

TRUSTED_SUSTAINABILITY

Specifications

Components PvuRts1I 5-hmC Endonuclease: 100U (1U/μL) in a buffer containing 20mM Tris pH 8.0, 150mM NaCl, 1mM DTT and 50% Glycerol. PvuRts1I 5-hmC 10X Reaction Buffer: 500μL of buffer containing 200mM Tris pH 8.0, 1.5M NaCl, 50mM MgCl2 and 10mM DTT.
For Use With (Application) Analyzing 5-hmC patterns genomic DNA by cleaving both glucosylated and non-glucosylated 5-hmC DNA sequences
Content And Storage -20°C for 12 months.
Quantity 200 U
Product Type PvuRts1I (5-hmC Restriction Endonuclease)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.