Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RAP, Human, Recombinant Protein, E. coli

Catalog No. 55350650UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-350-650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-350-650UG Supplier MilliporeSigma™ Supplier No. 55350650UG
Add to Cart
Add to Cart
Recombinant, human RAP fused to GST, expressed in E. coli, and subsequently cleaved with thrombin to remove GST. Antagonizes the ligand binding of certain members of the low-density lipoprotein receptor family such as low density lipoprotein receptor-related protein (LRP) and very low-density lipoprotein (VLDL) receptor. Promotes the proper folding and subsequent exocytic trafficking of the wild-type low density lipoprotein receptor (LDLR) and several of its class 2 mutants. Expressed as a GST-fusion protein that is subsequently cleaved with thrombin. Typical concentrations to block LRP or VLDL receptor are 200-300nM. For antagonism of LDLR, 1μM is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation 150mM NaCl, 100mM Tris-HCl, pH 7.4.
Purification Method SDS-PAGE
Quantity 50 μg
Source E. coli
Immunogen RAP
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.