Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ReBlot Plus Kit and Reagents
GSA_VA

Catalog No. 2504MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
Ea.
Product Type:
Mild WB Stripping Buffer
Strong WB Stripping Buffer
Western Blot Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
2504MI 50 mL Strong WB Stripping Buffer
2500MI Ea. Western Blot Kit
2502MI 50 mL Mild WB Stripping Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2504MI Supplier MilliporeSigma™ Supplier No. 2504
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Mild Antibody Stripping Solution (10X), 50mL; Strong Antibody Stripping Solution (10X), 50mL

Blots can be stripped of antibodies in approximately 15 minutes at room temperature

Western blotting is a commonly used technique for studying protein function and localization. Typically, protein samples are electrophoresed on SDS-PAGE and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid-based technologies, which allow reuse of Southern and Northern blots, it has been difficult to reuse Western blots.

  • No pungent-smelling β-mercaptoethanol is contained in the Antibody Stripping Solution
  • Antibody stripping is done at room temperature—no heating of blots is required
  • Blots may be reused in 25 min.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Strong WB Stripping Buffer
For Use With (Application) Western Blot
Quantity 50 mL
Concentration 2X
Content And Storage 50mL Strong Antibody stripping solution 10X. 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.