Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 50mM sodium acetate, 5mM EDTA, 1mM benzamidine, pH 6.0. |
| Purification Method | NuPAGE gel |
| Quantity | 150 mIU |
| Immunogen | Renin |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.