Learn More
MilliporeSigma™ Restriction Grade Factor Xa
Catalog No. 690363 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
69-036-3 100 μg
1 options
Catalog No. 69-036-3 Supplier MilliporeSigma™ Supplier No. 690363
Add to Cart
Add to Cart
a highly purified preparation isolated from bovine plasma and activated with Russell's viper venom.
This preparation is purified to near homogeneity and shows no secondary cleavage from contaminating proteases. The preparation is also functionally tested for activity with fusion proteins. Like enterokinase, Factor Xa cleaves at the C-terminal side of the recognition sequence (IleGluGlyArg↓) and can therefore be used for removing all vector-encoded sequences from appropriately designed constructs.