Learn More
MilliporeSigma™ RoClean™ Cleaning Pouches for Water Purification Systems
Catalog No. ZWACID012 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Acid Cleaning Pouch
Base Cleaning Pouch
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
ZWACID012 Acid Cleaning Pouch
ZWBASE012 Base Cleaning Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ZWACID012 Supplier MilliporeSigma™ Supplier No. ZWACID012
Keep your purification system clean with easy-to-use RoClean pouches.
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
†For details, see Catalog 2004/05.
TRUSTED_SUSTAINABILITY
Specifications
Type Acid Cleaning Pouch
For Use With (Equipment) Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system