Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SeQuant™ ZIC™-HILIC HPLC Guard Columns

Catalog No. M1504920001 Shop All MilliporeSigma Products
Change view
Click to view available options
Length (Metric):
5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
M1504920001 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1504920001 Supplier MilliporeSigma™ Supplier No. 1504920001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

guard column

SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. These guard columns protect your capillary separation column from contamination.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 300 μm
Length (Metric) 5 mm
Particle Size 5 μm
Pore Size 200 Å
Porosity 200 Å
Quantity 5/Pk.
Stationary Phase Sulfobetaine and phosphorylcholine
Product Line ZIC-HILIC
Phase HILIC
Material PEEK
Technique HILIC
Product Type Guard Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.