Learn More
MilliporeSigma™ SNAP i.d. 2.0™ Double MultiBlot Protein Detection System
Catalog No. SNAP2MB3 Shop All MilliporeSigma Products
Change view
Click to view available options
Dimensions (L x W):
4.5 x 8.4 cm (1.7 x 3.3 in.)
Catalog No. Dimensions (L x W)
SNAP2MB3 4.5 x 8.4 cm (1.7 x 3.3 in.)
1 options
Catalog No. SNAP2MB3 Supplier MilliporeSigma™ Supplier No. SNAP2MB3
Add to Cart
Add to Cart
For use on immunoblots
This system includes the SNAP i.d. 2.0™ Base, two MultiBlot Holding Frames, two Antibody Collection Trays, vacuum tubing, Rolling Pad & Blot Roller, two Wetting Trays and a Quick Start Guide
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot
Quantity 1/Pk.
Dimensions (L x W) 4.5 x 8.4 cm (1.7 x 3.3 in.)