Learn More
MilliporeSigma™ SNAP i.d.™ 2.0 MultiBlot Holding Frames and Holders
Catalog No. p-4842413 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
MultiBlot Holder
MultiBlot Holding Frame
Quantity:
50/Pk.
Single Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
SNP2BHMB050 MultiBlot Holder 50/Pk.
SNAP2FRMB01 MultiBlot Holding Frame Single Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SNP2BHMB050 Supplier MilliporeSigma™ Supplier No. SNAP2BHMB050
Only null left
Add to Cart
Add to Cart
For SNAP i.d.™ 2.0 System
Multiblot Holder
  • Each half blot membrane is aligned horizontally, allowing for uniform distribution of antibody
  • Flat profile allows for compact space allocation for storage, shipping and waste management
  • Size: 4.5 x 8.4cm
Multiblot Holding frame
  • Stackable frame with lid, easy locking latch and umbrella valve for offline incubation
  • Clear lids retains humidity
Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Type MultiBlot Holder
Quantity 50/Pk.