Learn More
MilliporeSigma™ SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames
Catalog No. p-4889740 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Midi Blot Holding Frames
Mini Blot Holding Frames
Quantity:
1/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
SNAP2FRMD01 Midi Blot Holding Frames 1/Pk.
SNAP2FRMN01 Mini Blot Holding Frames 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SNAP2FRMD01 Supplier MilliporeSigma™ Supplier No. SNAP2FRMD01
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mini and midi blot holding frames for SNAP i.d. 2.0 Protein Detection Systems
For use with EMD Millipore SNAP i.d. 2.0 Protein Detection System (Cat. No. SNAP2MM)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Snap i.d.™ 2.0 Protein Detection System-midi
Type Midi Blot Holding Frames
Includes Clear lids, easy locking latch, umbrella valve
Quantity 1/Pk.
Stackable Yes