Learn More
MilliporeSigma™ SNAP i.d.™ Protein Detection System: Blot Holders
Catalog No. WBAVDBH022 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Double Well Blot Holder
Quantity:
60/Pk.
Device Holds:
4.6 x 8.8 cm Blots
Catalog No. Type Quantity Device Holds
WBAVDBH022 Double Well Blot Holder 60/Pk. 4.6 x 8.8 cm Blots
1 options
Catalog No. WBAVDBH022 Supplier MilliporeSigma™ Supplier No. WBAVDBH022
Blot holders for use with SNAP i.d. Protein Detection System
TRUSTED_SUSTAINABILITY
Specifications
Device Holds 4.6 x 8.8 cm Blots
For Use With (Application) Western Blot
For Use With (Equipment) Snap i.d.™ Protein Detection System
Type Double Well Blot Holder
Includes Spacers
No. of Wells 2
Quantity 60/Pk.