Learn More
MilliporeSigma™ Sphingolipid Ceramide N-Deacylase, Pseudomonas sp., Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 50mM sodium acetate buffer, 0.1% LUBROL™ PX Detergent, pH 6.0. |
| Quantity | 250 mIU |
| Source | Pseudomonas sp. |
| Immunogen | Sphingolipid Ceramide N-Deacylase |
| Storage Requirements | Following initial thaw, aliquot and freeze (−20°C). |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.