Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | In 250mM phosphate buffer, 50% glycerol, pH 7.5. |
| Quantity | 10 U |
| Source | Staphylococcus aureus |
| Immunogen | Sphingomyelinase |
| Storage Requirements | Following initial use, aliquot and refrigerate (4°C). |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.