Learn More
MilliporeSigma™ Accessories for Sterifil™ Aseptic Systems
Catalog No. XX1104706 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Base and Support Screen
Receiver Flask Cover
Sterifil Holder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
XX1104706 Receiver Flask Cover
XX11J4750 Sterifil Holder
XX1104702 Base and Support Screen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. XX1104706 Supplier MilliporeSigma™ Supplier No. XX1104706
Accessories for Sterifil™ Aseptic Systems used for general filtration applications and filtration of samples for particulate or biological contamination analysis.
MilliporeSigma Polypropylene Support Screen, For Use With: For Swinnex 47, Surface Area: Support Screen, 47mm dia. Filter, Polypropylene, for Swinnex 47
  • For use with Sterifil™ Aseptic Systems.
Order Info
The Sterifil holder and funnel available separately (without receiver flask and cover) to use with standard 1 L vacuum filtering flask or multiple place manifold.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Sterifil Aseptic System
Material Polysulfone
Sterility Sterile
Type Receiver Flask Cover
Diameter (English) Inside 1.85 in.
Diameter (Metric) Inside 47 mm