Learn More
Description
Includes
6mm (0.25 in. ) stepped hose barb inlet, 6mm (0.25 in. ) stepped hose barb outlet and filling bell.
Fast flow and high throughput
Steripak™-GP filter units are designed for larger scale, pressure-driven filtration of tissue culture media with or without serum. Each filter is for a single use and is not autoclavable. The filter units come in two sizes with the smaller unit able to filter up to 10 L and the larger up to 20 L.
Features & Benefits:
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding
- Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
- 10 L and 20 L capacity sizes for maximum laboratory-scale filtration options
- Manually operated top air vent to prevent air lock stop of flow
- Filling bell for cleaner filling environment around the opening of the receptacle bottle
Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation
Specifications
Specifications
| Connectors | 6 mm stepped hose barb |
| Disposable | Single-use |
| Filter Type | Express PLUS |
| Material | SAN |
| Material (Membrane) | PES |
| Pore Size | 0.22 μm |
| Sterility | Sterile |
| Filtration Area | 200 cm2 |
| For Use With (Application) | Large-scale, pressure-driven tissue culture media filtration with or without serum |
| Max. Inlet Pressure | 75 psi forward, 5 psi reverse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.