Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Superoxide Dismutase, Human, Recombinant Protein, E. coli

Catalog No. 574591100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-459-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-459-1100UG Supplier MilliporeSigma™ Supplier No. 574591100UG
Add to Cart
Add to Cart
Recombinant, human superoxide dismutase expressed in E. coli as a homodimer consisting of 2 identical, 154-amino acid chains. Purified by proprietary chromatographic methods. Superoxide dismutase is an essential component of biological defense against the toxic effects of oxygen. It catalyzes the dismutation of superoxide radicals to molecular oxygen and H2O2 and plays a role in cellular aging due to its reduced activity in aging cells. Mutant forms of Cu/Zn SOD are associated with neurodegenerative disease.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM NaCl, 50mM sodium phosphate buffer.
Preparation Method Reconstitute in sterile MQ water to a final concentration of ≥100μg/mL. Further dilute in aqueous buffers just prior to use.
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Immunogen Superoxide Dismutase
Storage Requirements Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°C or for up to 4 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Solid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.