Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α-Synuclein, Human, Recombinant Protein, E. coli

Catalog No. 575001200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-500-1200UG 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-500-1200UG Supplier MilliporeSigma™ Supplier No. 575001200UG
Add to Cart
Add to Cart
Recombinant, human α-synuclein expressed in E. coli. α-Synuclein is an acidic neuronal protein of 140 amino acids (amino acid residues 1 -140). This enzyme has been implicated in the pathogenesis of Parkinson's disease and related neurodegenerative disorders, and more recently, to be an important regulatory component of vesicular transport in neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 20mM Tris-HCl, 1mM MgCl2, pH 7.5.
Purification Method SDS-PAGE
Quantity 200 μg
Source E. coli
Immunogen α-Synuclein
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.