Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TACE, HisTag™, Human, Recombinant Protein, T. ni.

Catalog No. PF13310UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF13310UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF13310UG Supplier MilliporeSigma™ Supplier No. PF13310UG
Add to Cart
Add to Cart
Recombinant, human TACE (amino acids 1-671) fused to a His·Tag™ sequence and expressed in T. ni. insect cells. The major form of the purified protein corresponds to the mature form with an apparent molecular mass of ∽70kDa.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered 25mM Tris-HCl, pH 8.0.
Preparation Method Reconstitute with 100μL dH2O.
Purification Method SDS-PAGE
Quantity 10 μg
Source T. ni.
Immunogen TACE
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Avoid repeated freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.