Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TdT, Calf Thymus, Protein

Catalog No. PF060155ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.155 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF060155ML 0.155 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF060155ML Supplier MilliporeSigma™ Supplier No. PF060.155ML
Add to Cart
Add to Cart
Native TdT from calf thymus. Tdt is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3' hydroxy terminus of either double of single stranded DNA in a template-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) DNA Labeling
Formulation 775 Units in 50mM potassium phosphate buffer, 1mM β-mercaptoethanol, 50% glycerol, pH 7.2.
Quantity 0.155 mL
Immunogen TdT
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.