Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thioredoxin Reductase, Yeast, Recombinant Protein, E. coli

Catalog No. 598502100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-850-2100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-850-2100UG Supplier MilliporeSigma™ Supplier No. 598502100UG
Add to Cart
Add to Cart
Recombinant, yeast thioredoxin reductase expressed in E. coli. Yeast thioredoxin reductase is a flavoprotein that has no selenocysteine (SeCys). It catalyzes the NADPH-dependent reduction of thioredoxin as well as other endogenous and exogenous proteins. Plays an important role in many aspects of cell function, including protection against oxidative stress/injury, cell growth and transformation, and the recycling of ascorbate from its oxidized form.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 150mM NaCl, 20mM phosphate buffer, pH 7.4.
Preparation Method Aqueous buffers, pH 7.0
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Immunogen Thioredoxin Reductase
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for 1 month at 4°C or 3 months at −70°C.
Regulatory Status RUO
Species Yeast
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.