Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thrombin, Bovine, High Activity, Protein

Catalog No. 604980100U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-498-0100U 100 U
60-498-0500U 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-498-0100U Supplier MilliporeSigma™ Supplier No. 604980100U
Add to Cart
Add to Cart
Native, bovine thrombin. Key enzyme that activates factor XIII and converts fibronogen into fibrin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 200mM NaCl, 50mM sodium citrate, 1mL 0.1% PEG, pH 6.5.
Preparation Method H2O
Purification Method Complete activation from homogeneous prothrombin by SDS-PAGE
Quantity 100 U
Immunogen Thrombin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.