Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 200mM NaCl, 50mM sodium citrate, 0.1% PEG-8000, pH 6.5. |
| Preparation Method | H2O (1mg/mL) |
| Purification Method | Complete activation from homogeneous bovine prothrombin by SDS-PAGE |
| Quantity | 1 kU |
| Immunogen | Thrombin |
| Storage Requirements | Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 6 months at −70°C. |
| Regulatory Status | RUO |
| Without Additives | Plasminogen-Free |
| Species | Bovine |
| Recombinant | Native |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.