Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thyroglobulin, Human Thyroid, Protein

Catalog No. 6093121MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-931-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-931-21MG Supplier MilliporeSigma™ Supplier No. 6093121MG
Add to Cart
Add to Cart
Native thyroglobulin, consisting of two polypeptide monomers of 33kDa each, purified from human thyroid. This protein is an excellent source of N-linked glycans of various structures, including sialylated core fucose and oligomannose oligosaccharides. Also, Gal-3-SO4 and GalNAc-6-SO4 residues located in a single multi-branched complex type glycan have been reported in calf thymus and human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 20mM NH4HCO3.
Preparation Method Reconstitute in phosphate buffer, containing 150mM NaCl, pH >7.0.
Purification Method PAGE
Quantity 1 mg
Immunogen Thyroglobulin
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.