Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tissue Factor, Apo-, Human, Recombinant Protein, S. frugiperda

Catalog No. 61215125UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-215-125UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-215-125UG Supplier MilliporeSigma™ Supplier No. 61215125UG
Add to Cart
Add to Cart
Recombinant, human apo-tissue factor expressed in S. frugiperda insect cells using a baculovirus expression system. Cell-surface protein of endothelial cells and monocytes involved in coagulation and inflammation. Primary cellular initiator of coagulation protease cascade. Functions as receptor and cofactor for coagulation Factors VII and VIIa. Requires lipidation for clotting assays or other functional activities.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Solid lyophilized from 200mM mannitol, 150mM NaCl, 10mM Tris-HCl, 0.01% CHAPS, pH 8.0.
Preparation Method dH2O
Purification Method SDS-PAGE
Quantity 25 μg
Source S. frugiperda
Immunogen Tissue Factor, Apo-
Storage Requirements Following reconstitution, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.