Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tissue Plasminogen Activator, Human, Protein

Catalog No. 612200100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-220-0100UG 100 μg
61-220-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-220-0100UG Supplier MilliporeSigma™ Supplier No. 612200100UG
Add to Cart
Add to Cart
Native tissue plasminogen activator from a human melanoma cell line. A serine protease that contains a single chain of 527 amino acids. Binds to fibrin via lysine binding sites at its amino terminus and activates bound plasminogen. Cleaves plasminogen to form active plasmin, the proteolytic enzyme that dissolves clots. Reported to be involved in neurite outgrowth, regeneration, and migration. Plays an important role in excitotoxin-induced neuronal degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM PBS, 3.5mg/mL L-arginine, 0.001% Tween™-80 Detergent.
Preparation Method Reconstitute with diH2O to a final concentration of 1mg/mL (e.g., 100μg should be reconstituted in 100μL).
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Tissue Plasminogen Activator
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.