Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TNF-α, Human, Recombinant Protein, E. coli

Catalog No. 65420510UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-420-510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-420-510UG Supplier MilliporeSigma™ Supplier No. 65420510UG
Add to Cart
Add to Cart
Recombinant, human tumor necrosis factor-α expressed in E. coli. A cytokine produced primarily by activated macrophages that activates a variety of immune defense mechanisms by interactions with polymorphonuclear leukocytes, T cells, antibody-producing B lymphocytes, fibroblasts, and hematopoietic bone marrow cells. Activity is not species-specific. Induces apoptosis in human blood and bone marrow neutrophils and in endothelial cells. Increases the iNOS levels in vascular smooth muscle cells. Involved in pathophysiological processes of several chronic and acute diseases. Stimulates stress activated protein kinase (SAPK).
TRUSTED_SUSTAINABILITY

Specifications

Formulation Carrier-free.
Preparation Method Reconstitute with sterile, dH2O to a final concentration of 0.1-1mg/mL. Further dilutions should be made in low endotoxin medium or aqueous buffer containing carrier protein such as heat inactivated FCS or tissue culture grade BSA.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen TNF-α
Storage Requirements Following reconstitution, aliquot and freeze (−20°C or −70°C) for long-term storage or refrigerate (4°C) for short-term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at −20°C or −70°C or up to 1 month at 4°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.