Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TRAIL, Human, Recombinant Protein, NSO cells

Catalog No. 61637610UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-637-610UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-637-610UG Supplier MilliporeSigma™ Supplier No. 61637610UG
Add to Cart
Add to Cart
Recombinant, human TRAIL expressed in NSO cells a type II transmembrane protein with a C-terminal extracellular domain, which exhibits homology to other TNF family members. Displays potent antitumor activity against selected targets. Induces apoptosis in a number of transformed cell lines but not in normal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS, 50μg BSA/μg TRAIL, pH 7.4.
Preparation Method Reconstitute to ≥20μg/mL in sterile PBS, 0.1% BSA.
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen TRAIL
Storage Requirements Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C or −70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or up to 3 months at −20°C or −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Cell Type NSO cells
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.