Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Transferrin Apo-, Human Plasma, Low Endotoxin Level, Protein

Catalog No. 61641910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-641-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-641-910MG Supplier MilliporeSigma™ Supplier No. 61641910MG
Add to Cart
Add to Cart
Native apotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free. Cell culture-tested.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 10mM phosphate buffer, pH 7.4.
Preparation Method H2O (10mg/mL)
Purification Method SDS-PAGE
Quantity 10 mg
Immunogen Transferrin, Apo-
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 year at −20°C.
Regulatory Status RUO
Endotoxin Concentration Low Endotoxin Level
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.