Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Transferrin, Apo-, Human Plasma, Protein

Catalog No. 616395100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-639-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-639-5100MG Supplier MilliporeSigma™ Supplier No. 616395100MG
Add to Cart
Add to Cart
Native apo-transferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 10mM sodium phosphate buffer, pH 7.4.
Preparation Method H2O (10mg/mL)
Purification Method SDS-PAGE
Quantity 100 mg
Immunogen Transferrin, Apo-
Storage Requirements Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 2 weeks at 4°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.