Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Transferrin, Holo, Human Plasma, Protein

Catalog No. 616397100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-639-7100MG 100 mg
61-639-7500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-639-7100MG Supplier MilliporeSigma™ Supplier No. 616397100MG
Add to Cart
Add to Cart
Native holotransferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 150mM NaCl, 20mM sodium phosphate buffer, pH 7.4.
Preparation Method H2O (10mg/mL)
Purification Method SDS-PAGE
Quantity 100 mg
Immunogen Transferrin, Holo
Storage Requirements Following reconstitution, aliquot and freeze (−20°C) for long-term storage or add NaN3 and refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.