Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Trypsin 0.25%

Catalog No. SM2001C Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SM2001C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SM2001C Supplier MilliporeSigma™ Supplier No. SM2001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Trypsin 0.25%, In Hank's Balanced Salt Solution, without Ca 2+ and Mg 2+ is available in a 100mL format.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20° C for 18 months
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.