Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ubiquitin, GST-Fusion, Recombinant Protein, E. coli

Catalog No. 6620571MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-205-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-205-71MG Supplier MilliporeSigma™ Supplier No. 6620571MG
Recombinant ubiquitin fused at the N-terminus to GST and expressed in E. coli. Useful for ubiquitination of protein substrates and subsequent glutathione affinity purification of ubiquitinated molecules. Can also be used for immunodetection of conjugates using GST antibodies. Facilitates the visualization of polyUb chains due to larger ladder intervals.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 50mM HEPES, pH 8.0.
Purification Method SDS-PAGE
Quantity 1 mg
Source E. coli
Immunogen Ubiquitin
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.