Learn More
MilliporeSigma™ Upstate™ CpGenome™ 5-mC and 5-hmC Human DNA Standard
Shop All MilliporeSigma ProductsDescription
- Matched genomic DNA standards originate from the same individual.
- Cytosine modifications are quantified by LC/MS for use as positive controls.
Specifications
Specifications
| Content And Storage | -20°C for 6 months |
| Description | CpGenome 5-mC and 5-hmC Human DNA Standard |
| Includes | Human spleen DNA, human brain DNA |
| Quantity | 5 μg/Vial |