Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CpGenome™ Human Methylated And Non-Methylated DNA Standard Set

Catalog No. S8001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S8001 5 μg/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8001 Supplier MilliporeSigma™ Supplier No. S8001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For gene methylation studies

  • Standards originate from DNA methyltransferase-knockout HCT116 cells.
  • Methylated control DNA is treated enzymatically at CpG dinucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C for 6 months
Description CpGenome human methylated and non-methylated DNA standard set
For Use With (Application) Gene methylation studies
Quantity 5 μg/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.