Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ CpGenome™ Human Methylated DNA Standard Set

Catalog No. S8001M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S8001M 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8001M Supplier MilliporeSigma™ Supplier No. S8001M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For gene methylation studies

  • Enzymatically methylated at CpG dinucleotides.
  • Derived from DNA methyltransferase-knockout cells.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C for 6 months
Description CpGenome human methylated DNA standard set
For Use With (Application) Gene methylation studies
Quantity 5 μg
Cell Line HCT-116
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.