Learn More
MilliporeSigma™ Upstate™ CpGenome™ Universal Methylated Mouse DNA Standard Set
Catalog No. S8000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
Catalog No. Quantity
S8000 5 μg
1 options
Catalog No. S8000 Supplier MilliporeSigma™ Supplier No. S8000
Add to Cart
Add to Cart
Contain universally methylated mouse genomic DNA isolated from male Balb/c mice, enzymatically methylated by methyltransferase
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C for 6 months
Description CpGenome universal methylated mouse DNA standard set
Quantity 5 μg