Learn More
MilliporeSigma™ Upstate H2A.X Phosphorylation Assay Kit
Catalog No. 17344 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Assays
Catalog No. Quantity
17-344 100 Assays
1 options
Catalog No. 17-344 Supplier MilliporeSigma™ Supplier No. 17344
Add to Cart
Add to Cart
For flow cytometric detection of levels of phosphorylated Histone H2A.X.
TRUSTED_SUSTAINABILITY
Specifications
Description H2A.X Phosphorylation Assay Kit
For Use With (Application) For flow cytometric detection of levels of phosphorylated Histone H2A.X.
Includes 10X Wash Solution, Anti-phospho H2A.X, FITC conjugate, Normal Mouse IgG, FITC conjugate, 16X Fixation Solution, 10X Permeabilization Solution
Quantity 100 Assays