Learn More
MilliporeSigma™ Upstate™ H2A.X Phosphorylation Assay Kit (Chemiluminescence Detection)
Shop All MilliporeSigma ProductsDescription
- Cell-based ELISA format.
- Uses a phospho-specific H2A.X antibody and an HRP-conjugated secondary antibody.
Specifications
Specifications
| Description | H2A.X Phosphorylation Assay Kit |
| Detection Method | Chemiluminescence |
| For Use With (Application) | ELISA |
| Includes | Goat Anti-Mouse IgG, HRP conjugate, Anti-phospho-Histone H2A.X (Ser139), clone JBW301, TBS, 20X, 20% Tween-20 (v/v), 10% BSA in TBS (Blocking Buffer), LumiGLO Substrate Reagent A, LumiGLO Substrate Reagent B |
| Quantity | 1 Kit (192 Assays) |
| Type | Cell-Based Assay Kit |
| Cell Line | Any Adherent Cell Type |
| Druggable Target | Cell-based Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.