Learn More
Specifications
Specifications
| Description | Histone Deacetylase (HDAC) assay Kit |
| Detection Method | Radioactive |
| For Use With (Application) | Enzyme Assays |
| Includes | PCAF active, HeLa Nuclear Extract, Histone H4 Peptide, biotin conjugate, residues 2-24, HAT Assay Buffer, 5X, Sodium Butyrate, 5X HDAC Assay Buffer, Streptavidin Agarose |
| Quantity | 1 Kit (100 Assays) |
| Cell Line | HeLa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.