Learn More
Specifications
Specifications
| Characterization | >90% by SDS-PAGE |
| For Use With (Application) | Enzyme Assay (HDAC deacetylase assays) |
| Formulation | In liquid containing 50mM Tris, pH 8.0, 138mM NaCl and 10% glycerol |
| Name | HDAC2 (1-488) (His-tag) |
| Quantity | 50 μg |
| Source | Insect cells |
| Storage Requirements | -80deg;C. |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.