Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Human Recombinant HDAC1 (Hist-tag)

Catalog No. 03190MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-190MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-190MI Supplier MilliporeSigma™ Supplier No. 03190
Add to Cart
Add to Cart

Specifications

Characterization >50% by SDS-PAGE
For Use With (Application) Enzyme Assay (HDAC deacetylase assays)
Formulation Liquid containing 50mM Tris, pH 8.0, 138mM NaCl and 10% glycerol
Name HDAC1 (Hist-tag)
Purification Method SDS-PAGE
Quantity 50 μg
Source Insect cells
Storage Requirements -80deg;C
Conjugate His-Tag
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.