Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Human Recombinant HDAC8

Catalog No. 03186MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-186MI 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-186MI Supplier MilliporeSigma™ Supplier No. 03186
Add to Cart
Add to Cart

Specifications

Characterization >50% by SDS-PAGE
For Use With (Application) Enzyme Assay (HDAC deacetylase assays)
Formulation Liquid containing 10mM Tris, pH 7.5, 100mM NaCl, 3mM MgCl2 and 10% glycerol
Name HDAC8
Purification Method SDS-PAGE
Quantity 100 U
Source E. coli
Storage Requirements -80deg;C
Cross Reactivity Human
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.